missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Attune™ Acoustic Focusing Cytometer Auto Sampler, without computer

Product Code. 13395384 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 Each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13395384 1 Each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13395384 Supplier Applied Biosystems™ Supplier No. 4473928

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

Attune™ Acoustic Focusing Cytometer Auto Sampler, without computer

The Attune™ Acoustic Focusing Cytometer Auto Sampler, without computer, is an add-on accessory compatible with those Attune™ Acoustic Focusing Cytometer models designed for use with this device (catalog numbers 4445315AS and 4469120AS). This auto sampler does not include a computer but does include an Attune™ Auto Sampler Accessory Kit. The Attune™ auto sampler enables rapid processing of multiple samples by automatically analyzing samples from 96- or 384-well standard and deep-well plates.

Some features of the Attune™ auto sampler are:

  • Flexible—accommodates multiple plate formats
  • Reliable—instrument design results in higher productivity and less down time
  • Accurate—sample mixing leads to consistent results
  • Simple to use—experiment setup has never been this simple

    Flexible
    The Attune™ auto sampler allows use of many different plate formats, including 96- and 384-well standard and deep-well plates, without being restricted to a single manufacturer. To maintain system flexibility, the manual sample probe is always accessible even when the loader is present, and switching between tubes and plates is as simple as a flip of a switch.

    Reliable
    The Attune™ auto sampler was designed to minimize system down time. Automated sampling devices are often plagued by clogging or probe crashes. The Intelliprobe™, with a 200 micron diameter that is larger than most probes available in the industry, minimizes instances of clogging.

    To prevent probe crashes, which may require expensive probe replacement, the Attune™ auto sampler automatically performs a probe self-calibration every month to ensure consistent positioning of the probe within every well. The self-calibration verifies proper function of the probe, reduces variability in probe positioning over time to prevent failures during plate runs, and improves reliability over the life of the system. Self-calibration of the probe also allows the auto sampler to be easily moved, so it can be shared between multiple instruments.

    In addition, the Attune™ auto sampler initiates automated cleaning procedures when the instrument is shutting down to ensure proper maintenance with little user interaction.

    Accurate
    Unlike other plate loaders that mix samples by shaking, which may not ensure homogeneity of the sample, the Attune™ auto sampler mixes samples by aspiration, ensuring uniform mixing while preserving sample viability and resulting in consistent data throughout the plate.

    Simple to Use
    The instrument design and software were developed with customer ease-of-use in mind. The Attune™ auto sampler has fluidic bottles with sensors that inform the user when the fluids need to be replaced. The auto sampler’s indicator lights and warning systems have been designed to ensure that a user who has walked away from the system can easily see the system status at all times.

    Setting up an experiment with the Attune™ cytometric software is simple. Using a virtual plate layout view, users can create compensation wells, define instrument settings, and identify samples. The software also allows multiple experiments to be defined on a single plate. The intuitive software allows quick learning and data analysis. For example, a heat map view allows rapid screening and confirmation of samples while the instrument is acquiring data.

    The software is free and downloadable online at www.thermofisher.com/attune. Multiple users can access the software. No license fees or dongles are required.

    For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Room temperature
    Includes Attune™ auto sampler, Attune™ auto sampler accessory kit
    Quantity 1 Each
    For Use With (Equipment) Attune Acoustic Focusing Cytometer
    Product Type Attune™ Acoustic Focusing Cytometer Auto Sampler

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.