Learn More
Invitrogen™ Attractin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532667
Description
Percent identity with other species by BLAST analysis: Human, Gibbon (100%); Monkey, Marmoset, Mouse, Rat, Dog, Hamster, Rabbit (93%); Gorilla, Bovine, Panda (87%); Bat, Pig (80%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Multiple transcript variants encoding different isoforms exist for this gene. One of the isoforms is a membrane-bound protein with sequence similarity to the mouse mahogany protein, a receptor involved in controlling obesity. The other isoform is a secreted protein involved in the initial immune cell clustering during inflammatory responses that may regulate the chemotactic activity of chemokines.
Specifications
| Attractin | |
| Polyclonal | |
| Unconjugated | |
| ATRN | |
| ATRN; atrn {ECO:0000312; Attractin; attractin-2; AW558010; DPPT-L; KIAA0548; mahogany; mahogany homolog; mahogany protein; membrane attractin; mg; MGC126754; Mgca; mKIAA0548; Protein mahogany; protein zitter; RGD:69063} | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8455 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O75882 | |
| ATRN | |
| Synthetic 15 amino acid peptide from internal region of human ATRN / Attractin. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.