Learn More
Scientific Industries Attachable 2nd Tier Platform
Product Code. 17251762 Shop All Ohaus Products
Change view
Click to view available options
No. of Positions:
1
Unit Size:
Each
Product Code. No. of Positions Unit Size
17251762 1 Each
1 options
Product Code. 17251762 Supplier Scientific Industries Supplier No. 83041265
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Attachable 2nd Tier Platform doubles your shaking capacity and is easily attached and secured without any tools. For use on the Mini-300 Orbital-Genies or any Genie Temp-Shaker models with Adhering Mat Platform or Flask Clamp Platform.
  • Attachable 2nd Tier Platform doubles your shaking capacity
  • Easily attached and secured without any tools
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vessel Platform
Weight (Metric) 1.81 kg
For Use With (Equipment) Mini-300 Orbital-Genies/Any Genie Temp-Shaker Models with Adhering Mat Platform/Flask Clamp Platform
No. of Positions 1
Warranty and Service Offering 30 Days