missing translation for 'onlineSavingsMsg'
Learn More

Atropine, TRC

Product Code. 30041260
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041260 1 g 1g
30041130 5 g 5g
30040851 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041260 Supplier Toronto Research Chemicals Supplier No. TRCA7946301G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-55-8
Molecular Formula C17 H23 N O3
Molecular Weight (g/mol) 289.37
Synonym 1αH,5αH-Tropan-3α-ol (±)-tropate (ester) (8CI), Benzeneacetic acid, α-(hydroxymethyl)-, 8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester, endo-, (3-endo)-8-Methyl-8-azabicyclo[3.2.1]oct-3-yl α-(hydroxymethyl)benzeneacetate, (±)-Atropine, (±)-Hyoscyamine, Atropin, Atropine, Atropinum sulphuricum, DL-Hyoscyamine, Tropine (±)-tropate, Tropine tropate, dl-Hyoscyamine, dl-Tropyl tropate
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2R)-3-hydroxy-2-phenylpropanoate
SMILES CN1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@@H](CO)c3ccccc3

Specifications

Analyte or Component Names Atropine
Shipping Condition Room Temperature
InChI Formula InChI=1S/C17H23NO3/c1-18-13-7-8-14(18)10-15(9-13)21-17(20)16(11-19)12-5-3-2-4-6-12/h2-6,13-16,19H,7-11H2,1H3/t13-,14+,15+,16-/m0/s1
Solution Type Neat
Industry Type Forensics
Quantity 1 g
Formula Weight 289.1678
Percent Purity >95%
Chemical Name or Material Atropine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.