missing translation for 'onlineSavingsMsg'
Learn More

Atropine sulfate monohydrate, MedChemExpress

Product Code. 16418286
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16418286 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418286 Supplier Medchem Express Supplier No. HYB0394
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Atropine (Tropine tropate) sulfate monohydrate is a broad-spectrum and competitive muscarinic acetylcholine receptor (mAChR) antagonist with anti-myopia effect.

Atropine (Tropine tropate) sulfate monohydrate is a broad-spectrum and competitive muscarinic acetylcholine receptor (mAChR) antagonist with anti-myopia effect.

Specifications

Chemical Name or Material Atropine sulfate monohydrate
Physical Form Powder
CAS 5908-99-6
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C17H23NO3.1/2H2O4S.1/2H2O
For Use With (Application) Neuroscience-Neuromodulation
Synonym Tropine tropate sulfate monohydrateDL-Hyoscyamine sulfate monohydrate
Solubility Information DMSO : 50 mg/mL (143.91 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (95.93 mM; Need ultrasonic)
SMILES O=[S](O)(O)=O.O=C(C(C1=CC=CC=C1)CO)O[C@H]2C[C@@H](CC3)N(C)[C@@H]3C2.[1/2].O.[1/2]
Molecular Weight (g/mol) 347.43
Formula Weight 347.43
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.