missing translation for 'onlineSavingsMsg'
Learn More

Atropine sulfate monohydrate, 97+%

Product Code. 11483933
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11483933

Brand: Thermo Scientific Alfa Aesar A10236.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5908-99-6 | C17H27NO8S | 405.462 g/mol

Antagonist at muscarinic receptors; also a bronchodilator. It is also used in used in preanesthetic medication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antagonist at muscarinic receptors; also a bronchodilator. It is also used in used in preanesthetic medication.

Solubility
Soluble in water (2500 mg/ml) at 4°C, ethanol (200 mg/ml) at 4°C, Glycerol (400 mg/ml), and methanol.

Notes
Light Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5908-99-6
Molecular Formula C17H27NO8S
Molecular Weight (g/mol) 405.462
MDL Number MFCD00074815
InChI Key PVGPXGKNDGTPTD-IJTOKZDFSA-N
Synonym atropine sulfate hydrate, atropine sulfate monohydrate, 1r,3s,5s-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 3-hydroxy-2-phenylpropanoate; methane; sulfuric acid
PubChem CID 23624044
IUPAC Name [(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] 3-hydroxy-2-phenylpropanoate;sulfuric acid;hydrate
SMILES CN1C2CCC1CC(C2)OC(=O)C(CO)C3=CC=CC=C3.O.OS(=O)(=O)O

Specifications

Melting Point 187°C to 192°C
Quantity 50 g
UN Number UN1544
Beilstein 6109275
Sensitivity Light sensitive
Merck Index 14,875
Solubility Information Soluble in water (2500mg/ml) at 4°C,ethanol (200mg/ml) at 4°C,Glycerol (400mg/ml),and methanol.
Formula Weight 694.84 (676.82 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material Atropine sulfate monohydrate
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.