missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atropine sulfate monohydrate, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Antagonist at muscarinic receptors; also a bronchodilator. It is also used in used in preanesthetic medication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Antagonist at muscarinic receptors; also a bronchodilator. It is also used in used in preanesthetic medication.
Solubility
Soluble in water (2500 mg/ml) at 4°C, ethanol (200 mg/ml) at 4°C, Glycerol (400 mg/ml), and methanol.
Notes
Light Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Antagonist at muscarinic receptors; also a bronchodilator. It is also used in used in preanesthetic medication.
Solubility
Soluble in water (2500 mg/ml) at 4°C, ethanol (200 mg/ml) at 4°C, Glycerol (400 mg/ml), and methanol.
Notes
Light Sensitive Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 187°C to 192°C |
Quantity | 10 g |
UN Number | UN1544 |
Beilstein | 6109275 |
Sensitivity | Light sensitive |
Merck Index | 14,875 |
Solubility Information | Soluble in water (2500mg/ml) at 4°C,ethanol (200mg/ml) at 4°C,Glycerol (400mg/ml),and methanol. |
Formula Weight | 694.84 (676.82 Anhydrous) |
Percent Purity | ≥98% |
Chemical Name or Material | Atropine sulfate monohydrate |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 2
- H300-Fatal if swallowed.
- H330-Fatal if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'