missing translation for 'onlineSavingsMsg'
Learn More

Atropine, MedChemExpress

Product Code. 16498676
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498676 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16498676 Supplier Medchem Express Supplier No. HYB1205
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Atropine (Tropine tropate) is a competitive muscarinic acetylcholine receptor (mAChR) antagonist with IC50 values of 0.39 and 0.71 nM for Human mAChR M4 and Chicken mAChR M4, respectively. Atropine inhibits ACh-induced relaxations in human pulmonary veins. Atropine can be used for research of anti-myopia and bradycardia.

Atropine (Tropine tropate) is a competitive muscarinic acetylcholine receptor (mAChR) antagonist with IC50 values of 0.39 and 0.71 nM for Human mAChR M4 and Chicken mAChR M4, respectively. Atropine inhibits ACh-induced relaxations in human pulmonary veins. Atropine can be used for research of anti-myopia and bradycardia.

Specifications

Chemical Name or Material Atropine
CAS 51-55-8
Color White
Purity Grade Notes Research
Molecular Formula C17H23NO3
Quantity 100 mg
Synonym Tropine tropate DL-Hyoscyamine
Solubility Information DMSO : ≥ 96.6 mg/mL (333.83 mM) ∣H2O : 2.9 mg/mL (10.02 mM; Need ultrasonic and warming)
SMILES O=C(O[C@@H]1C[C@@H](N2C)CC[C@@H]2C1)C(CO)C3=CC=CC=C3
Molecular Weight (g/mol) 289.37
Formula Weight 289.37
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.