missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATRIP Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA596590

Product Code. 16519431

  • £439.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAGTSAPGSK RRSEPPAPRP GPPPGTGHPP SKRARGFSAA AAPDPDDPFG AHGDFTADDL EELDTLASQA LSQCPAAARD VSSDHKVHRL LDGMSKNPSG KNRETVPIKD NFELEVLQAQ YKELKEKMKV MEEEVLIKNG EIKILRDSLH QTESVLEEQR RSHFLLEQEK TQALSDKEKE FSKKLQSLQS ELQFKDAEMN ELRTKLQTSE RANKLAAPSV SHVSPRKNPS VVIKPEACSP QFGKTSFPTK ESFSANMSLP; Positive Samples: HepG2, Jurkat, HeLa, Mouse brain, Mouse heart; Cellular Location: Nucleus.

ATRIP is an 86 kDa protein that is phosphorylated by ATR, regulates ATR expression, and is an essential component of the DNA damage checkpoint pathway. It localizes to the intranuclear foci after DNA damage or inhibition of replication.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATRIP
Polyclonal
Unconjugated
ATRIP
6620401K05Rik; AGS1; ATM and Rad3 related interacting protein; ATM and Rad3-related-interacting protein; ATR interacting protein; ATR IP; ATR-interacting protein; Atrip; DNase III; DRN3; FLJ12343; unnamed protein product
Rabbit
Affinity chromatography
RUO
235610, 84126
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.87 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BMG1, Q8WXE1
ATRIP
Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human ATRIP (NP_5690551).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.