missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ATRIP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29219
This item is not returnable.
View return policy
Description
ATRIP Polyclonal antibody specifically detects ATRIP in Human samples. It is validated for Immunohistochemistry
Specifications
| ATRIP | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| Unconjugated | |
| AGS1, ATM and Rad3-related-interacting protein, ATR interacting protein, ATR-interacting protein, DKFZp762J2115, FLJ12343, MGC20625, MGC21482, MGC26740 | |
| Between 100 and 150 | |
| 25 μg | |
| Cancer, Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair | |
| 84126 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction