missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Atrial Natriuretic Peptide (ANP) Competitive ELISA Kit

Product Code. 15845853
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
Pack of 10
Pack of 96
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15845853 96 Tests Pack of 96
17825117 10 x 96 Tests Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15845853 Supplier Invitrogen™ Supplier No. EIAANP
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

ELISA

The Atrial Natriuretic Peptide (ANP) Competitive ELISA quantitates ANP in human plasma,urine or cell culture medium.

Principle of the method

The Atrial Natriuretic Peptide (ANP) Competitive ELISA research-use-only kit is designed to quantitatively measure ANP present in plasma,urine,and tissue culture media samples. This kit is not recommended for serum or plasma samples without extraction. An ANP standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture rabbit antibodies. An ANP-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of an antibody to ANP to each well. After a 1-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound ANP-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.

Rigorous validation

Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene belongs to the natriuretic peptide family. Natriuretic peptides are implicated in the control of extracellular fluid volume and electrolyte homeostasis. This protein is synthesized as a large precursor (containing a signal peptide), which is processed to release a peptide from the N-terminus with similarity to vasoactive peptide, cardiodilatin, and another peptide from the C-terminus with natriuretic-diuretic activity. Mutations in this gene have been associated with atrial fibrillation familial type 6.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.741 to 180 ng/mL
Assay Sensitivity 0.25 ng/mL
Description Atrial Natriuretic Peptide (ANP) Competitive ELISA Kit
Assay Multiplex assay on the Luminex Platform
Conjugate HRP
Detection Method Fluorescence
Product Type ELISA
Protein Family Peptide hormone
Sample Type Plasma, Supernatant, Urine
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 7.6%
Intraassay CV 6.7
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Inflammation
Sample Volume Plasma 50 μL, Supernatant 50 μL, Urine 12.5 μL
Storage Requirements -20°C
Target ANP
Test Time 1 hrs 20 mins
Total Assay Time 1 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.