missing translation for 'onlineSavingsMsg'
Learn More

Atrial Natriuretic Peptide/ANP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-35467-20ul

 View more versions of this product

Product Code. 30228499

  • £145.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Atrial Natriuretic Peptide/ANP Polyclonal antibody specifically detects Atrial Natriuretic Peptide/ANP in Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Atrial Natriuretic Peptide/ANP
Polyclonal
Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
ANF, ANPatriopeptin, ATFB6, atrial natriuretic factor, Atrial natriuretic peptide, CDD-ANF, natriuretic peptide A, natriuretic peptide precursor A, PNDcardionatrin, prepronatriodilatin
Recombinant fusion protein containing a sequence corresponding to amino acids 26-151 of human Atrial Natriuretic Peptide/ANP (NP_006154.1).,, Sequence:, LTWQEIMSITELQGLNAPSEPSFEPQAPAPYLGPPPPTTYCPCSIHPDSGFPLPPPPYELPASTSHVPDPPYSYGNMAIPVSKPLSLSGLLSEPLQDPLALLDIGLPAGPPKPQEDPESDSGLSLN
20 μL
Primary
Mouse, Rat
Purified
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
4878
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.