missing translation for 'onlineSavingsMsg'
Learn More

Atrial Natriuretic Peptide/ANP Antibody (2E3), Novus Biologicals™

Product Code. p-200022554 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30025270 50 μL 50µL
30024421 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30025270 Supplier Novus Biologicals Supplier No. NBP32646050ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Atrial Natriuretic Peptide/ANP Monoclonal antibody specifically detects Atrial Natriuretic Peptide/ANP in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Atrial Natriuretic Peptide/ANP
Applications ELISA
Classification Monoclonal
Clone 2E3
Conjugate Unconjugated
Dilution ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias ANF, ANPatriopeptin, ATFB6, atrial natriuretic factor, Atrial natriuretic peptide, CDD-ANF, natriuretic peptide A, natriuretic peptide precursor A, PNDcardionatrin, prepronatriodilatin
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Atrial Natriuretic Peptide/ANP [UniProt P01160]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4878
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.