missing translation for 'onlineSavingsMsg'
Learn More

Atractyligenin, TRC

Product Code. 30334336
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30334336 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334336 Supplier Toronto Research Chemicals Supplier No. TRCA7945251MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Atractyligenin
CAS 10391-47-6
Molecular Formula C19H28O4
Quantity 1 mg
Synonym 2beta,15alpha-Dihydroxy-19-norkaur-16-en-18-oic Acid, (2beta,4alpha,15alpha)-2,15-Dihydroxy-19-norkaur-16-en-18-oic Acid, 1H-2,10a-Ethanophenanthrene, 19-norkaur-16-en-18-oic Acid deriv., (4alpha)-2beta,15alpha-Dihydroxy-19-norkaur-16-en-18-oic Acid
InChI Formula InChI=1S/C19H28O4/c1-10-11-3-4-15-18(2)9-12(20)7-13(17(22)23)14(18)5-6-19(15,8-11)16(10)21/h11-16,20-21H,1,3-9H2,2H3,(H,22,23)/t11-,12-,13-,14-,15+,16+,18-,19-/m1/s1
SMILES C[C@@]12C[C@H](O)C[C@H]([C@H]1CC[C@@]34C[C@@H](CC[C@@H]23)C(=C)[C@@H]4O)C(=O)O
IUPAC Name (1R,4R,5R,7R,9R,10S,13R,15S)-7,15-dihydroxy-9-methyl-14-methylidenetetracyclo[11.2.1.01,10.04,9]hexadecane-5-carboxylic acid
Molecular Weight (g/mol) 320.42
Formula Weight 320.1988
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.