missing translation for 'onlineSavingsMsg'
Learn More

Atractylenolide III, MedChemExpress

Product Code. 16498888
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16498888

Brand: Medchem Express HYN0203

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Atractylenolide III is a major component of Atractylodes rhizome can induce apoptosis of the lung carcinoma cells.

Atractylenolide III is a major component of Atractylodes rhizome can induce apoptosis of the lung carcinoma cells.

Specifications

Chemical Name or Material Atractylenolide III
CAS 73030-71-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H20O3
Quantity 10 mg
Synonym ICodonolactone 8β-Hydroxyasterolide
Solubility Information DMSO : 50 mg/mL (201.35 mM; Need ultrasonic)
SMILES O=C1C(C)=C(C[C@@]23[H])[C@@](C[C@@]3(C)CCCC2=C)(O)O1
Molecular Weight (g/mol) 248.32
Formula Weight 248.32
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.