missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ATPgammaS tetralithium salt
GREENER_CHOICE

Product Code. 18094552
Change view
Click to view available options
:
10mg
Product Code.
18094552 10mg
1 options
This item is not returnable. View return policy

Product Code. 18094552

Brand: Tocris Bioscience™ 4080/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Non-selective P2 agonist; analog of ATP (Cat. No. 3245)

ATPγS tetralithium salt is a P2 purinergic receptor agonist. Nonhydrolyzable analog of ATP (Cat. No. 3245).
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type ATPγS tetralithium salt
Purity >90%
Molecular Weight (g/mol) 546.98

Product is intended for laboratory research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.