missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATPG Antibody (OTI2H6), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70233PE
This item is not returnable.
View return policy
Description
ATPG Monoclonal antibody specifically detects ATPG in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| ATPG | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI2H6 | |
| Western Blot, Immunohistochemistry | |
| ATP synthase gamma chain, mitochondrial, ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1, ATP5CATP synthase subunit gamma, mitochondrial, ATP5CL1mitochondrial ATP synthase, gamma subunit 1, F-ATPase gamma subunit | |
| Full length human recombinant protein of human ATP5C1 (NP_001001973) produced in E.coli. | |
| 0.1 mL | |
| Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 509 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction