missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Highly selective compound, ubiquinone-binding site mitochondrial complex II inhibitor, stimulates mitochondrial channels
- Shows anti-ischemic and cardioprotective effects in vivo
- Used in the Krebs cycle to fumarate with the reduction of quinone to quinol
Specifications
Specifications
| CAS | 119509-24-9 |
| Synonym | FO 125A5, atpenin A5, Antibiotic FO 125A5, Atpenin A5 (Synthetic), (2 S,4 S,5 R)-5,6-Dichloro-1-(2,4-dihydroxy-5,6-dimethoxypyridin-3-yl)-2,4-dimethyl-1-hexanone, (2 S,4 S,5 R)-1-(2,4-Dihydroxy-5,6-dimethoxy-3-pyridinyl)-2,4-dimethyl-5,6-dichloro-1-hexanone, 3-[(2 S,4 S,5 R)-5,6-Dichloro-2,4-dimethyl-1-oxohexyl]-4-hydroxy-5,6-dimethoxy-2(1 H)-pyridinone |
| Quantity | 1 mg |
| Melting Point | 83°C to 85°C |
| Shipping Condition | -20°C |
| Molecular Formula | C15H21Cl2NO5 |
| Color | White |
| Purity | 96.1 |
| Molecular Weight (g/mol) | 366.2, 366.24 |
| InChI Key | OVULNOOPECCZRG-CIUDSAMLSA-N |
| Show More |
Safety and Handling
Product Identifier
- 5-Iodo-2'-deoxyuridine
- Danger
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1A
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H340-May cause genetic defects.
- H361-Suspected of damaging fertility or the unborn child.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 5-Iodo-2'-deoxyuridine
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?