missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATPBD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560181
This item is not returnable.
View return policy
Description
Immunogen sequence: ISGGKDSCYN MMQCIAAGHQ IVALANLRPA ENQVGSDELD SYMYQTVGHH AIDLYAEAMA LPLYRRTIRG RSLDTRQVYT KCE Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
Amidase that catalyzes the last step of diphthamide biosynthesis using ammonium and ATP. Diphthamide biosynthesis consists in the conversion of an L-histidine residue in the translation elongation factor (EEF2) to diphthamide.
Specifications
| ATPBD4 | |
| Polyclonal | |
| Unconjugated | |
| DPH6 | |
| 5730421E18Rik; ATP binding domain 4; Atpbd4; ATP-binding domain-containing protein 4; diphthamide synthase; Diphthamide synthetase; diphthamine biosynthesis 6; diphthine--ammonia ligase; DPH6; DPH6 homolog; protein DPH6 homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 89978 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7L8W6 | |
| DPH6 | |
| Recombinant protein corresponding to Human ATPBD4. Recombinant protein control fragment (Product #RP-98065). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction