Learn More
ATPBD4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565952
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: GDEVEDLYELL KLVKEKEEVE GISVGAILSD YQRIRVENVC KRLNLQPLAY LWQRNQEDLL REMISSNIQA MIIKVAALGL DPDKHLG Highest antigen sequence identity to the following orthologs - mouse 93%, rat 65%.
Amidase that catalyzes the last step of diphthamide biosynthesis using ammonium and ATP. Diphthamide biosynthesis consists in the conversion of an L-histidine residue in the translation elongation factor (EEF2) to diphthamide.Specifications
ATPBD4 | |
Polyclonal | |
Unconjugated | |
DPH6 | |
5730421E18Rik; ATP binding domain 4; Atpbd4; ATP-binding domain-containing protein 4; diphthamide synthase; Diphthamide synthetase; diphthamine biosynthesis 6; diphthine--ammonia ligase; DPH6; DPH6 homolog; protein DPH6 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
89978 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7L8W6 | |
DPH6 | |
Recombinant Human ATPBD4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only