missing translation for 'onlineSavingsMsg'
Learn More

ATPase Na+/K+ beta 2 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18433359 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433359 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433359 Supplier Novus Biologicals Supplier No. NBP297186IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATPase Na+/K+ beta 2 Polyclonal antibody specifically detects ATPase Na+/K+ beta 2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATPase Na+/K+ beta 2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias adhesion molecule on glia, AMOGsodium/potassium-dependent ATPase beta-2 subunit, ATPase, Na+/K+ transporting, beta 2 polypeptide, Na, K-ATPase beta-2 polypeptide, Sodium/potassium-dependent ATPase subunit beta-2, sodium/potassium-transporting ATPase beta-2 chain, sodium/potassium-transporting ATPase subunit beta-2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ATPase Na+/K+ beta 2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 482
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.