missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Atpalpha Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA532093

Product Code. 13416667

  • £442.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: Drosophila lysate. Predicted reactivity: Drosophila (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The ATPa, or ATPalpha gene, encodes a subunit of the Na+/K+ ATPase enzyme, which is crucial for maintaining cellular ion gradients by actively transporting Na+ out of cells and K+ into cells. This gene is essential in various organisms, including Drosophila melanogaster (fruit fly). Mutations in the ATPalpha gene can lead to significant developmental and physiological abnormalities, such as neurodegeneration, altered longevity, and myodegeneration. One of its critical roles is in neuromuscular functioning, where it affects locomotor activity, with mutations causing conditional bang-sensitive or temperature-sensitive paralysis. Additionally, ATPalpha mutations disrupt the formation and function of septate junctions in Drosophila, structures analogous to vertebrate tight junctions, further underscoring its role in maintaining cellular integrity and signal transduction.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Atpalpha
Polyclonal
Unconjugated
Atpalpha
(Na[+] + K[+])-ATPase alpha-subunit; (Na[+],K[+])ATPase alpha subunit; (Na[+]/k[+])-ATPase; alpha-ATPase; alpha-subunit of ATPase; Atp; Atp a; ATPa; atpalpha; ATP-alpha; Atpalpha-PA; Atpalpha-PB; Atpalpha-PC; Atpalpha-PD; Atpalpha-PE; Atpalpha-PF; Atpalpha-PG; Atpalpha-PH; Atpalpha-PI; Atpalpha-PJ; Atpalpha-PK; ATPase alpha; ATPase alpha subunit; ATPBagr; CG5670; CG5670-PA; CG5670-PB; CG5670-PC; CG5670-PD; CG5670-PE; CG5670-PF; CG5670-PG; CG5670-PH; CG5670-PI; CG5670-PJ; CG5670-PK; Da-47; Dmel\CG5670; Dmel_CG5670; dnATPase; JY; K ATPase; K[+] -ATPase; K[+]-ATPase; K[+]-ATPase alpha subunit; K[+]-ATPase alpha-subunit; K-ATPase; l(3)01164; l(3)01453; l(3)04492; l(3)j3B2; l(3)j3B7; l(3)j6A4; l(3)rM454; l(3)ry136; Na; Na pump alpha subunit; Na(+)/K(+) ATPase alpha subunit; Na, K ATPase a subunit; Na,K ATPase; Na,K ATPase alpha subunit; Na,K-ATPase; Na,K-ATPase alpha; Na,K-ATPase alpha subunit; Na,K-ATPase alpha-subunit; Na/K ATPase; Na/K-ATPase; Na/K-ATPase alpha-subunit; Na[ +], K[ +] -ATPase; Na[+]; Na[+] pump alpha subunit; Na[+], K[+] ATPase alpha; Na[+],K[+] ATPase alpha subunit; Na[+],K[+]-ATPase alpha subunit; Na[+]/K[+] ATPase; Na[+]/K[+] ATPase (alpha subunit); Na[+]/K[+] ATPase alpha subunit; Na[+]/K[+]ATPase; Na[+]/K[+]-ATPase; Na[+]K[+] ATPase; Na[+]-K[+] ATPase; Na[+]K[+] ATPase alpha subunit; Na[+]K[+]ATPase; Na[+]K[+]-ATPase; Na[+]-K[+]-ATPase alpha[[5]]-subunit; Na+,K+ ATPase alpha-subunit; Na+/K+ ATPase alpha subunit; Na+/K+-ATPase; Na+-K+-ATPase; NaK; NaK ATPase; NaK-ATPase; Na-p; NKA; sodium pump alpha subunit; sodium pump alpha-subunit; Sodium pump subunit alpha; sodium/potassium transporting ATPase alpha; Sodium/potassium-transporting ATPase subunit alpha
Rabbit
Antigen affinity chromatography
RUO
48971
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
P13607
Atpalpha
Recombinant protein encompassing a sequence within the center region of Drosophila Atpalpha. The exact sequence is proprietary.
100 μL
Primary
Drosophila, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.