missing translation for 'onlineSavingsMsg'
Learn More

ATP8B3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18379271
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379271 20 μg 20µL
18304554 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18379271 Supplier Bio-Techne Supplier No. NBP31555120UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP8B3 Polyclonal antibody specifically detects ATP8B3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP8B3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias probable phospholipid-transporting ATPase IK, ATPase, aminophospholipid transporter, class I, type 8B, member 3, ATPIK
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 600-900 of human ATP8B3 (NP_620168.1).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 148229
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.