missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP8B2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524267
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse based on sequence homology.
The protein encoded by this gene belongs to the family of P-type cation transport ATPases, and to the subfamily of aminophospholipid-transporting ATPases. The aminophospholipid translocases transport phosphatidylserine and phosphatidylethanolamine from one side of a bilayer to another. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| ATP8B2 | |
| Polyclonal | |
| Unconjugated | |
| ATP8B2 | |
| 36/8-9 fusion protein with epitope for anti-lectin antibody; ATP8B2; ATPase class I type 8B member 2; ATPase phospholipid transporting 8B2; ATPase, aminophospholipid transporter, class I, type 8B, member 2; ATPase, class I, type 8B, member 2; ATPID; KIAA1137; P4-ATPase flippase complex alpha subunit ATP8B2; phospholipid-transporting ATPase ID; probable phospholipid-transporting ATPase ID | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 57198 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P98198 | |
| ATP8B2 | |
| KLH conjugated synthetic peptide between 18-47 amino acids from the N-terminal region of human AT8B2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction