missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP8B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57019
This item is not returnable.
View return policy
Description
ATP8B2 Polyclonal specifically detects ATP8B2 in Human samples. It is validated for Western Blot.
Specifications
| ATP8B2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ATP8B2 | |
| Synthetic peptides corresponding to ATP8B2 (ATPase, class I, type 8B, member 2) The peptide sequence was selected from the N terminal of ATP8B2. Peptide sequence KTSKYNILTFLPVNLFEQFQEVANTYFLFLLILQLIPQISSLSWFTTIVP. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| ATPase, class I, type 8B, member 2, ATPIDprobable phospholipid-transporting ATPase ID, DKFZp434M0219, EC 3.6.3, EC 3.6.3.1, KIAA1137ATPase class I type 8B member 2, phospholipid-transporting ATPase ID | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57198 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction