missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP8A2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA565256

Product Code. 15958014

  • £476.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GVTYGHFPELA REPSSDDFCR MPPPCSDSCD FDDPRLLKNI EDRHPTAPCI QEFLTLLAVC HTVVPEKDGD N Highest antigen sequence identity to the following orthologs - mouse 88%, rat 89%.

Catalytic component of a P4-ATPase flippase complex which catalyzes the hydrolysis of ATP coupled to the transport of aminophospholipids from the outer to the inner leaflet of various membranes and ensures the maintenance of asymmetric distribution of phospholipids. Phospholipid translocation seems also to be implicated in vesicle formation and in uptake of lipid signaling molecules. Reconstituted to liposomes, the ATP8A2:TMEM30A flippase complex predomiminantly transports phosphatidylserine (PS) and to a lesser extent phosphatidylethanolamine (PE). Proposed to function in the generation and maintenance of phospholipid asymmetry in photoreceptor disk membranes and neuronal axon membranes. May be involved in vesicle trafficking in neuronal cells. Involved in regulation of neurite outgrowth; acting in synergy with TMEM30A. Required for normal visual and auditory function; involved in photoreceptor and inner ear spiral ganglion cell survival.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATP8A2
Polyclonal
Unconjugated
ATP8A2
ATP; ATP8A2; ATPase class I type 8A member 2; ATPase phospholipid transporting 8A2; ATPase, aminophospholipid transporter, class I, type 8A, member 2; ATPase, aminophospholipid transporter-like, class I, type 8A, member 2; ATPIB; CAMRQ4; IB; ML-1; P4-ATPase flippase complex alpha subunit ATP8A2; Phospholipid-transporting ATPase IB; probable phospholipid-transporting ATPase IB
Rabbit
Antigen affinity chromatography
RUO
51761
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NTI2
ATP8A2
Recombinant Human ATP8A2. Recombinant protein control fragment (Product #RP-105975).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.