missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP7A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103110
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ATP7A.
ATP7A (also known as Copper-transporting ATPase 1) functions as a transmembrane copper-translocating P-type ATPase and plays a vital role in systemic copper absorption in the gut and copper reabsorption in the kidney. Polarized epithelial cells such as Madin-Darby canine kidney cells are a physiologically relevant model for systemic copper absorption and reabsorption in vivo. Although ATP7A is not detectable in most normal tissues, it is expressed in a considerable fraction of many common tumor types. Increased expression of ATP7A renders cells resistant to cisplatin and carboplatin. Mutations in the ATP7A gene result in Menkes disease, which is fatal in early childhood.
Specifications
| ATP7A | |
| Polyclonal | |
| Unconjugated | |
| ATP7A | |
| ATP 7A; ATP7A; ATPase copper transporting alpha; ATPase, Cu++ transporting, alpha polypeptide; copper pump 1; Copper-transporting ATPase 1; Cu++-transporting P-type ATPase; DSMAX; FLJ17790; MC 1; MC1; Menke; Menkes disease-associated protein; menkes disease-associated protein homolog; Menkes protein; Menkes syndrome; MK; MNK; OHS; OTTHUMP00000062077; SMAX3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11977, 24941, 538 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P70705, Q04656, Q64430 | |
| ATP7A | |
| A synthesized peptide derived from human ATP7A(Accession Q04656), corresponding to amino acid residues S1463-L1500. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction