Learn More
ATP7A Mouse anti-Human, Rat, Clone: S60-4, Abnova™
Mouse monoclonal antibody raised against human ATP7A.
Brand: Abnova MAB12725.50ug
Description
This gene encodes a transmembrane protein that functions in copper transport across membranes. The protein localizes to the trans-Golgi network, where it is predicted to supply copper to copper-dependent enzymes in the secretory pathway. The protein relocalizes to the plasma membrane under conditions of elevated extracellular copper and functions in the efflux of copper from cells. Mutations in this gene result in Menkes disease, X-linked cutis laxa, and occipital horn syndrome. [provided by RefSeq]
Specifications
| ATP7A | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user. | |
| FLJ17790/MK/MNK | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 538 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b |
| Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| S60-4 | |
| Mouse monoclonal antibody raised against human ATP7A. | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
| ATP7A | |
| A synthetic peptide corresponding to amino acids 42-61 at cytoplasmic region of human ATP7A. | |
| 50 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.