missing translation for 'onlineSavingsMsg'
Learn More

ATP7A Antibody (S60-4), Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18430083 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430083 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430083 Supplier Novus Biologicals Supplier No. NBP259376JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ATP7A Monoclonal antibody specifically detects ATP7A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP7A
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone S60-4
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias ATPase, Cu++ transporting, alpha polypeptide, Copper pump 1, copper-transporting ATPase 1, Cu++-transporting P-type ATPase, DSMAX, EC 3.6.3, EC 3.6.3.4, MC1, Menkes disease-associated protein, Menkes syndrome, MK, MNKFLJ17790, OHS, SMAX3
Host Species Mouse
Immunogen Synthetic peptide amino acids 42-61 (cytoplasmic C-terminus) of human Copper- transporting ATPase1
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 538
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.