missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP7A Antibody (S60-4), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59376H
This item is not returnable.
View return policy
Description
ATP7A Monoclonal specifically detects ATP7A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ATP7A | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| ATP7A | |
| Synthetic peptide amino acids 42-61 (cytoplasmic C-terminus) of human Copper- transporting ATPase1 | |
| 100 μL | |
| 538 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| S60-4 | |
| Western Blot, Immunohistochemistry | |
| ATPase, Cu++ transporting, alpha polypeptide, Copper pump 1, copper-transporting ATPase 1, Cu++-transporting P-type ATPase, DSMAX, EC 3.6.3, EC 3.6.3.4, MC1, Menkes disease-associated protein, Menkes syndrome, MK, MNKFLJ17790, OHS, SMAX3 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects the extracellular domain of LRP4 215kDa. Degradation product can be seen at approx 150kDa. | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction