missing translation for 'onlineSavingsMsg'
Learn More

ATP7A Antibody, Novus Biologicals™

Product Code. 18324182 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324182 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324182 Supplier Novus Biologicals Supplier No. NBP311275

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP7A Polyclonal specifically detects ATP7A in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP7A
Applications Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias ATPase, Cu++ transporting, alpha polypeptide, Copper pump 1, copper-transporting ATPase 1, Cu++-transporting P-type ATPase, DSMAX, EC 3.6.3, EC 3.6.3.4, MC1, Menkes disease-associated protein, Menkes syndrome, MK, MNKFLJ17790, OHS, SMAX3
Host Species Rabbit
Immunogen Peptide (Asp1475-Leu1492) situated at the C-terminus of mouse (NM009726) and rat (NM052803) ATP7A
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 538
Target Species Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.