missing translation for 'onlineSavingsMsg'
Learn More

ATP6V1H Antibody, Novus Biologicals™

Product Code. 18397899 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397899 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397899 Supplier Novus Biologicals Supplier No. H00051606D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP6V1H Polyclonal antibody specifically detects ATP6V1H in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V1H
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_057025.2
Gene Alias ATPase, H+ transporting, lysosomal 50/57kD, V1 subunit H, ATPase, H+ transporting, lysosomal 50/57kDa, V1 subunit H, CGI-11, MSTP042, NBP1, Nef-binding protein 1, Protein VMA13 homolog, SFD, SFDalpha, SFDbeta, vacuolar ATP synthase subunit H, vacuolar ATPase subunit H, vacuolar proton pump H subunit, Vacuolar proton pump subunit H, Vacuolar proton pump subunit SFD, V-ATPase 50/57 kDa subunits, V-ATPase H subunit, V-ATPase subunit H, VMA13, V-type proton ATPase subunit H
Host Species Rabbit
Immunogen ATP6V1H (NP_057025.2, 1 a.a. - 483 a.a.) full-length human protein. MTKMDIRGAVDAAVPTNIIAAKAAEVRANKVNWQSYLQGQMISAEDCEFIQRFEMKRSPEEKQEMLQTEGSQCAKTFINLMTHICKEQTVQYILTMVDDMLQENHQRVSIFFDYARCSKNTAWPYFLPMLNRQDPFTVHMAARIIAKLAAWGKELMEGSDLNYYFNWIKTQLSSQKLRGSGVAVETGTVSSSDSSQYVQCVAGCLQLMLRVNEYRFAWVEADGVNCIMGVLSNKCGFQLQYQMIFSIWLLAFSPQMCEHLRRYNIIPVLSDILQESVKEKVTRIILAAFRNFLEKSTERETRQEYALAMIQCKVLKQLENLEQQKYDDEDISEDIKFLLEKLGESVQDLSSFDEYSSELKSGRLEWSPVHKSEKFWRENAVRLNEKNYELLKILTKLLEVSDDPQVLAVAAHDVGEYVRHYPRGKRVIEQLGGKQLVMNHMHHEDQQVRYNALLAVQKLMVHNWEYLGKQLQSEQPQTAAARS
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51606
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.