missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1G3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £359.00
Specifications
| Antigen | ATP6V1G3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661402
|
Novus Biologicals
NBP2-92742-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615802
|
Novus Biologicals
NBP2-92742-0.1ml |
0.1 mL |
£359.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATP6V1G3 Polyclonal antibody specifically detects ATP6V1G3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ATP6V1G3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 127124 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| ATPase, H+ transporting, lysosomal 13kD, V1 subunit G isoform 3, ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G isoform 3, ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G3, H+ transporting, lysosomal (vacuolar proton pump) subunit G3, MGC119810, MGC119813, vacuolar ATP synthase subunit G 3, vacuolar proton pump G subunit 3, Vacuolar proton pump subunit G 3, vacuolar proton pump, subunit G3, V-ATPase 13 kDa subunit 3, V-ATPase G subunit 3, V-ATPase G3 subunit, V-ATPase subunit G 3, V-type proton ATPase subunit G 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-118 of human ATP6V1G3 (NP_573569.1). MTSQSQGIHQLLQAEKRAKDKLEEAKKRKGKRLKQAKEEAMVEIDQYRMQRDKEFRLKQSKIMGSQNNLSDEIEEQTLGKIQELNGHYNKYMESVMNQLLSMVCDMKPEIHVNYRATN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title