missing translation for 'onlineSavingsMsg'
Learn More

ATP6V1G2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18381557 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381557 25 μg 25µL
18394247 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18381557 Supplier Novus Biologicals Supplier No. NBP31712125UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP6V1G2 Polyclonal antibody specifically detects ATP6V1G2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V1G2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias ATP6G2, ATPase, H+ transporting, lysosomal (vacuolar proton pump), ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G isoform 2, ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G2, Em:AC004181.3, subunit G2, vacuolar ATP synthase subunit G 2, vacuolar proton pump G subunit 2, Vacuolar proton pump subunit G 2, V-ATPase 13 kDa subunit 2, V-ATPase subunit G 2, Vma10, V-type proton ATPase subunit G 2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: NLSAEVEQATRRQVQGMQSSQQRNRERVLAQLLGMVC
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 534
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.