missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1F Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17725-1-AP-20UL
This item is not returnable.
View return policy
Description
ATP6V1F encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c′, c", and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is the V1 domain F subunit protein.Specifications
| ATP6V1F | |
| Polyclonal | |
| Unconjugated | |
| ATP6V1F | |
| ATP6S14, ATP6V1F, V ATPase 14 kDa subunit, V ATPase subunit F, V type proton ATPase subunit F, Vacuolar proton pump subunit F, VATF, Vma7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66144, 9296 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q16864, Q9D1K2 | |
| Atp6v1f | |
| ATP6V1F Fusion Protein Ag12121 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction