missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP6V1F Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563998
This item is not returnable.
View return policy
Description
Immunogen sequence: DIGIILINQY IAEMVRHALD AHQQSIPAVL EIPSKEHPYD AAKDSILRRA RGMFTAEDL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
ATP6V1F encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c', and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is the V1 domain F subunit protein.
Specifications
| ATP6V1F | |
| Polyclonal | |
| Unconjugated | |
| Atp6v1f | |
| 1110004G16Rik; adenosinetriphosphatase 14k chain; ATP6S14; ATP6V1F; ATPase H+ transporting V1 subunit F; ATPase, H transporting, lysosomal V1 subunit F; ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F; ATPase, H+ transporting, lysosomal V1 subunit F; ATPase, H+ transporting, V1 subunit F; ATPase, vacuolar, 14 kD; H(+)-transporting two-sector ATPase, 14kD subunit; vacuolar ATP synthase subunit F; vacuolar proton pump F subunit; vacuolar proton pump subunit F; VATF; V-ATPase 14 kDa subunit; V-ATPase F subunit; V-ATPase subunit F; Vma7; V-type proton ATPase subunit F; v-type proton ATPase subunit F-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9296 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16864 | |
| Atp6v1f | |
| Recombinant protein corresponding to Human V-ATPase F. Recombinant protein control fragment (Product #RP-103427). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction