missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ATP6V1E2 Recombinant Protein Antigen

Product Code. 18391225 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391225 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18391225 Supplier Novus Biologicals™ Supplier No. NBP317046PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 90423
Molecular Weight (g/mol) M.W. Theoretical: 20 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias ATP6E1V-ATPase subunit E 2, ATP6EL2, ATP6V1EL2, ATPase, H+ transporting, lysosomal (vacuolar proton pump) 31kD-like 2, ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E isoform 2, ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E2, MGC9341, Vacuolar proton pump subunit E 2, vacuolar-type proton-translocating ATPase subunit E1, VMA4, V-type proton ATPase subunit E 2
Gene Symbol ATP6V1E2
Research Category Cancer, Endocrinology, Signal Transduction
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein ATP6V1E2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.