missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1E2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-17046-100UL
This item is not returnable.
View return policy
Description
ATP6V1E2 Polyclonal antibody specifically detects ATP6V1E2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ATP6V1E2 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| ATP6E1V-ATPase subunit E 2, ATP6EL2, ATP6V1EL2, ATPase, H+ transporting, lysosomal (vacuolar proton pump) 31kD-like 2, ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E isoform 2, ATPase, H+ transporting, lysosomal 31kDa, V1 subunit E2, MGC9341, Vacuolar proton pump subunit E 2, vacuolar-type proton-translocating ATPase subunit E1, VMA4, V-type proton ATPase subunit E 2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: AKLRLSRIVEDPEVYQGLLDK | |
| 100 μg | |
| Cancer, Endocrinology, Signal Transduction | |
| 90423 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction