missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14920-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c′, c", and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene encodes the V1 domain D subunit protein.Specifications
| ATP6V1D | |
| Polyclonal | |
| Unconjugated | |
| ATP6V1D | |
| ATP6M, ATP6V1D, V ATPase subunit D, V type proton ATPase subunit D, Vacuolar proton pump subunit D, VATD, VMA8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 299159, 51382, 73834 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P57746, Q9Y5K8 | |
| ATP6V1D | |
| ATP6V1D Fusion Protein Ag6737 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction