missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1C2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16274-1-AP-150UL
This item is not returnable.
View return policy
Description
ATP6V1C2 encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A,three B, and two G subunits, as well as a C, D, E, F, and H subunit. The V1 domain contains the ATP catalytic site. This gene encodes alternate transcriptional splice variants, encoding different V1 domain C subunit isoforms.Specifications
| ATP6V1C2 | |
| Polyclonal | |
| Unconjugated | |
| ATP6V1C2 | |
| ATP6C2, ATP6V1C2, V ATPase subunit C 2, VMA5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 245973, 68775 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8NEY4, Q99L60 | |
| Atp6v1c2 | |
| ATP6V1C2 Fusion Protein Ag9349 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction