missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1C2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33020
This item is not returnable.
View return policy
Description
ATP6V1C2 Polyclonal specifically detects ATP6V1C2 in Human samples. It is validated for Western Blot.
Specifications
| ATP6V1C2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| ATP6V1C2 | |
| Recombinant protein encompassing a sequence within the center region of human V-ATPase C2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 245973 | |
| Human | |
| IgG |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| ATP6C2, ATPase, H+ transporting, lysosomal 42kD, V1 subunit C isoform 2, ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C isoform 2, ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C2, Vacuolar proton pump subunit C 2, V-ATPase C2 subunit, V-ATPase subunit C 2, VMA5, V-type proton ATPase subunit C 2 | |
| Rabbit | |
| 49 kDa | |
| 100 μL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction