missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP6V1A Recombinant Rabbit Monoclonal Antibody (24GB6860)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556988
This item is not returnable.
View return policy
Description
ATP6V1A Recombinant Monoclonal Antibody for Western Blot
ATP6V1A encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c', and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is one of two V1 domain A subunit isoforms and is found in all tissues. Transcript variants derived from alternative polyadenylation exist.
Specifications
| ATP6V1A | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ATP6V1A | |
| 70-kDa subunit; AI647066; Atp6a1; Atp6a2; Atp6v1a; Atp6v1a1; ATPase H+ transporting V1 subunit A; ATPase, H+ transporting, lysosomal (vacuolar proton pump), alpha 70 kDa, isoform 2; ATPase, H+ transporting, lysosomal 70kD, V1 subunit A, isoform 1; ATPase, H+ transporting, lysosomal 70kDa, V1 subunit A; ATPase, H+ transporting, lysosomal V1 subunit A; ATPase, H+ transporting, lysosomal, subunit A1; ATPase, H+ transporting, V1 subunit A, isoform 1; ATPase, H+ transporting, V1 subunit A1; H(+)-transporting two-sector ATPase, subunit A; H+-transporting ATPase chain A, vacuolar (VA68 type); HO68; lysosomal 70kDa; VA68; vacuolar ATP synthase catalytic subunit A, ubiquitous isoform; vacuolar ATPase isoform VA68; vacuolar proton pump alpha subunit 1; vacuolar proton pump subunit alpha; V-ATPase 69 kDa subunit; V-ATPase 69 kDa subunit 1; V-ATPase A subunit 1; V-ATPase subunit A; Vma1; VPP2; V-type proton ATPase catalytic subunit A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11964, 523, 685232 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB6860 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P38606, P50516 | |
| ATP6V1A | |
| Recombinant protein of human ATP6V1A (aa 62-78). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction