missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ATP6V1A |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ATP6V1A Polyclonal specifically detects ATP6V1A in Human samples. It is validated for Western Blot.Specifications
| ATP6V1A | |
| Polyclonal | |
| Rabbit | |
| P38606 | |
| 523 | |
| Synthetic peptides corresponding to ATP6V1A(ATPase, H+ transporting, lysosomal 70kDa, V1 subunit A) The peptide sequence was selected from the N terminal of ATP6V1A. Peptide sequence SGVSVGDPVLRTGKPLSVELGPGIMGAIFDGIQRPLSDISSQTQSIYIPR. | |
| Primary | |
| 68 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 70kD, isoform 1, ATP6A1, ATP6V1A1ATPase, H+ transporting, lysosomal, subunit A1, ATPase, H+ transporting, lysosomal (vacuolar proton pump), alpha polypeptide, ATPase, H+ transporting, lysosomal 70kDa, V1 subunit A, EC 3.6.3, EC 3.6.3.14, H(+)-transporting two-sector ATPase, subunit A, H+-transporting ATPase chain A, vacuolar (VA68 type), HO68, VA68, vacuolar ATP synthase catalytic subunit A, ubiquitous isoform, Vacuolar ATPase isoform VA68, vacuolar proton pump alpha subunit 1, Vacuolar proton pump subunit alpha, V-ATPase 69 kDa subunit, V-ATPase 69 kDa subunit 1, V-ATPase A subunit 1, V-ATPase subunit A, Vma1, VPP2, V-type proton ATPase catalytic subunit A | |
| ATP6V1A | |
| IgG | |
| This product is specific to Subunit or Isoform: A. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title