missing translation for 'onlineSavingsMsg'
Learn More

ATP6V1A Antibody (GT1561), Novus Biologicals™

Product Code. 18399566 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399566 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399566 Supplier Novus Biologicals Supplier No. NBP313542

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ATP6V1A Monoclonal antibody specifically detects ATP6V1A in Human samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V1A
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone GT1561
Conjugate Unconjugated
Formulation PBS
Gene Alias 70kD, isoform 1, ATP6A1, ATP6V1A1ATPase, H+ transporting, lysosomal, subunit A1, ATPase, H+ transporting, lysosomal (vacuolar proton pump), alpha polypeptide, ATPase, H+ transporting, lysosomal 70kDa, V1 subunit A, EC 3.6.3, EC 3.6.3.14, H(+)-transporting two-sector ATPase, subunit A, H+-transporting ATPase chain A, vacuolar (VA68 type), HO68, VA68, vacuolar ATP synthase catalytic subunit A, ubiquitous isoform, Vacuolar ATPase isoform VA68, vacuolar proton pump alpha subunit 1, Vacuolar proton pump subunit alpha, V-ATPase 69 kDa subunit, V-ATPase 69 kDa subunit 1, V-ATPase A subunit 1, V-ATPase subunit A, Vma1, VPP2, V-type proton ATPase catalytic subunit A
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human ATP6V1A. The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 523
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.