missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATP6V0E2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569806
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100% This antibody recognizes isoforms 2 and 3 of human vacuolar ATPase.
Multisubunit vacuolar-type proton pumps, or H(+)-ATPases, acidify various intracellular compartments, such as vacuoles, clathrin-coated and synaptic vesicles, endosomes, lysosomes, and chromaffin granules. H(+)-ATPases are also found in plasma membranes of specialized cells, where they play roles in urinary acidification, bone resorption, and sperm maturation. Multiple subunits form H(+)-ATPases, with proteins of the V1 class hydrolyzing ATP for energy to transport H+, and proteins of the V0 class forming an integral membrane domain through which H+ is transported. ATP6V0E2 encodes an isoform of the H(+)-ATPase V0 e subunit, an essential proton pump component.
Specifications
| ATP6V0E2 | |
| Polyclonal | |
| Unconjugated | |
| ATP6V0E2 | |
| 0610006O14Rik; ATP6V0E2; ATP6V0E2L; ATPase H+ transporting V0 subunit e2; ATPase, H+ transporting V0 subunit e2; ATPase, H+ transporting, lysosomal V0 subunit E2; ATPase, H+ transporting, lysosomal, V0 subunit E; ATPase, H+ transporting, V0 subunit E; C7orf32; H+-ATPase e2 subunit; lysosomal 9 kDa H(+)-transporting ATPase V0 subunit e2; NM9.2; similar to ATPase, H+ transporting, V0 subunit; vacuolar proton pump subunit e 2; vacuolar proton-ATPase subunit; V-ATPase subunit e 2; V-type proton ATPase subunit e 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 155066 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NHE4 | |
| ATP6V0E2 | |
| synthetic peptide directed towards the middle region of human ATP6V0E2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction