missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V0D2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54595
This item is not returnable.
View return policy
Description
ATP6V0D2 Polyclonal specifically detects ATP6V0D2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| ATP6V0D2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ATP6D2, ATP6V0, ATPase, H+ transporting, lysosomal 38kD, V0 subunit d isoform 2, ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d isoform 2, ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d2, FLJ38708, Vacuolar proton pump subunit d 2, V-ATPase subunit d 2, VMA6, V-type proton ATPase subunit d 2 | |
| Rabbit | |
| 40 kDa | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: d 2. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8N8Y2 | |
| ATP6V0D2 | |
| Synthetic peptides corresponding to ATP6V0D2(ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d2) The peptide sequence was selected from the middle region of ATP6V0D2. Peptide sequence GLRLLAQAEDFDQMKNVADHYGVYKPLFEAVGGSGGKTLEDVFYEREVQM | |
| Affinity purified | |
| RUO | |
| 245972 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction