missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ATP6V0D1 Recombinant Rabbit Monoclonal Antibody (24GB6855)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen ATP6V0D1
Clone 24GB6855
Concentration 0.7 mg/mL
Content And Storage -20°C
Applications Flow Cytometry, Western Blot, Immunocytochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30276577
GREENER_CHOICE
View Documents
Invitrogen™
MA556985
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276659
View Documents
Invitrogen™
MA556986
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ATP6V0D1 Recombinant Monoclonal Antibody for Western Blot

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, , and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is known as the D subunit and is found ubiquitously.
Specifications

Specifications

ATP6V0D1
0.7 mg/mL
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P51863, P61421
11972, 291969, 9114
Recombinant protein of human ATP6V0D1 (aa 1-351).
Antibody
24GB6855
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
ATP6V0D1
32 kd accessory protein; 32 kDa accessory protein; Ac39; AI267038; Atp6d; ATP6DV; Atp6v0d1; ATPase H+ transporting V0 subunit d1; ATPase, H+ transporting, lysosomal (vacuolar proton pump), 42 kDa; ATPase, H+ transporting, lysosomal (vacuolar proton pump), member D; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit D; ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d1; ATPase, H+ transporting, lysosomal V0 subunit D1; ATPase, H+ transporting, lysosomal, V0 subunit d1; ATPase, H+ transporting, V0 subunit D; cto; fb73h07; FLJ43534; H(+)-transporting two-sector ATPase, subunit D; lysosomal 38kDa; P39; Physophilin; Unknown (protein for MGC:63769); Vacuolar proton pump subunit d 1; vacuolar proton-ATPase, subunit D; V-ATPase, subunit D; V-ATPase 40 KDa accessory protein; V-ATPase AC39 subunit; V-ATPase subunit d 1; V-ATPase, subunit D; VATX; Vma6; VPATPD; V-type proton ATPa; V-type proton ATPase subunit d 1; wu:fb73h07; zgc:63769
ATP6V0D1
Primary
Affinity chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.