missing translation for 'onlineSavingsMsg'
Learn More

ATP6V0D1 Antibody (3B8), Novus Biologicals™

Product Code. 18378238 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18378238 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18378238 Supplier Novus Biologicals Supplier No. H00009114M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ATP6V0D1 Monoclonal antibody specifically detects ATP6V0D1 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V0D1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3B8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_004682
Gene Alias 32 kDa accessory protein, ATP6D, ATP6DV, ATP6V0, ATPase, H+ transporting, lysosomal (vacuolar proton pump), member D, ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d isoform 1, ATPase, H+ transporting, lysosomal 38kDa, V0 subunit d1, FLJ43534, H(+)-transporting two-sector ATPase, subunit D, P39, Vacuolar proton pump subunit d 1, V-ATPase 40 kDa accessory protein, V-ATPase AC39 subunit, V-ATPase, subunit D, VATX, Vma6, VPATPDV-ATPase subunit d 1, V-type proton ATPase subunit d 1
Host Species Mouse
Immunogen ATP6V0D1 (NP_004682, 238 a.a. ~ 308 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. AKLFPHCGRLYPEGLAQLARADDYEQVKNVADYYPEYKLLFEGAGSNPGDKTLEDRFFEHEVKLNKLAFLN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9114
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.