missing translation for 'onlineSavingsMsg'
Learn More

ATP6V0A4 Antibody, DyLight 755, Novus Biologicals™

Product Code. 18329182 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329182 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329182 Supplier Novus Biologicals Supplier No. NBP299250IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP6V0A4 Polyclonal specifically detects ATP6V0A4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6V0A4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 755
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias A4, ATP6N2, ATP6V0, ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 1B, ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 2 (38kD), ATPase, H+ transporting, lysosomal V0 subunit a4, MGC130016, MGC130017, noncatalytic accessory protein 1B, RDRTA2, RTA1C, RTADR, STV1, vacuolar proton pump 116 kDa accessory subunit, vacuolar proton pump, subunit 2, vacuolar proton translocating ATPase 116 kDa subunit a kidney isoform, V-ATPase 116 kDa, VPH1, VPP2, V-type proton ATPase 116 kDa subunit a, V-type proton ATPase 116 kDa subunit a isoform 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human ATP6V0A4.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50617
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.