missing translation for 'onlineSavingsMsg'
Learn More

ATP6V0A4 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-92499-0.1ml

 View more versions of this product

Product Code. 18636222

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

ATP6V0A4 Polyclonal antibody specifically detects ATP6V0A4 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ATP6V0A4
Polyclonal
Western Blot 1:500-1:2000
A4, ATP6N2, ATP6V0, ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 1B, ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 2 (38kD), ATPase, H+ transporting, lysosomal V0 subunit a4, MGC130016, MGC130017, noncatalytic accessory protein 1B, RDRTA2, RTA1C, RTADR, STV1, vacuolar proton pump 116 kDa accessory subunit, vacuolar proton pump, subunit 2, vacuolar proton translocating ATPase 116 kDa subunit a kidney isoform, V-ATPase 116 kDa, VPH1, VPP2, V-type proton ATPase 116 kDa subunit a, V-type proton ATPase 116 kDa subunit a isoform 4
Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human ATP6V0A4 (NP_065683.2). MVSVFRSEEMCLSQLFLQVEAAYCCVAELGELGLVQFKDLNMNVNSFQRKFVNEVRRCESLERILRFLEDEMQNEIVVQLLEKSPLTPLPREMITLETVLEKLEGELQEANQNQQALKQSFLELTELKYLLKKTQDFFETETNLADDFFTEDTSGLLELKAVPAYMTGKLGFIAGVINRERMASFERLLWRICRGNVYLKFSEMDAPLEDPVTKEEIQKNIFIIFYQGEQLRQKIKKICDGFRATVYPCPEPAVERREMLESVNVRLEDLITVITQTESH
0.1 mL
Cancer, Endocrinology, Neuroscience, Signal Transduction
50617
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.